Läs mer
Flavanthrone, TRC
Produktkod. 30352799
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352799 100 mg
30361852 50 mg
30360051 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352799 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC382253100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 475-71-8
Molekylformel C28H12N2O2
Molekylvikt (g/mol) 408.41
Synonym Benzo[h]benz[5,6]acridino[2,1,9,8-klmna]acridine-8,16-dione
IUPAC-namn 16,30-diazaoctacyclo[15.11.1.14,28.02,15.03,12.05,10.018,23.025,29]triaconta-1(28),2(15),3(12),4(30),5,7,9,13,16,18,20,22,25(29),26-tetradecaene-11,24-dione
LEDER O=C1c2ccccc2c3nc4ccc5C(=O)c6ccccc6c7nc8ccc1c3c8c4c57
Specifikationer
Kemiskt namn eller material Flavanthrone
Kvantitet 100 mg
InChI formel InChI=1S/C28H12N2O2/c31-27-15-7-3-1-5-13(15)25-21-17(27)9-12-20-23(21)24-19(29-25)11-10-18-22(24)26(30-20)14-6-2-4-8-16(14)28(18)32/h1-12H
Formel vikt 408.0899