Läs mer
Flecainide Meta-O-dealkylated, TRC
Product Code. 30404669
Change view
Click to view available options
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30404669 25 mg
30396284 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30404669 Supplier Toronto Research Chemicals Supplier No. TRCF39000525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83526-33-4
Molekylformel C15H19F3N2O3
Molekylvikt (g/mol) 332.32
Synonym S 24623, 5-Hydroxy-N-(2-piperidinylmethyl)-2-(2,2,2-trifluoroethoxy)benzamide, 5-O-(Detrifluoroethane) Flecainide, 5-Hydroxy-N-(piperidin-2-ylmethyl)-2-(2,2,2-trifluoroethoxy)benzamide
LEDER Oc1ccc(OCC(F)(F)F)c(c1)C(=O)NCC2CCCCN2
Specifications
Kemiskt namn eller material Flecainide Meta-O-dealkylated
Kvantitet 25 mg
InChI formel InChI=1S/C15H19F3N2O3/c16-15(17,18)9-23-13-5-4-11(21)7-12(13)14(22)20-8-10-3-1-2-6-19-10/h4-5,7,10,19,21H,1-3,6,8-9H2,(H,20,22)
Formel vikt 332.1348