Läs mer
OHAUS™ Flex System Clamp
Handla allt Ohaus Produkter
Längd (metrisk) arm:
305 mm
457 mm
Inkluderar:
Base Plate
Lab Frame Clamp
Shelf Clamp
Förpackningsstorlek:
Varje
Beskrivning
- Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
- Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
- Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
Specifikationer
Specifikationer
| Typ | Flex System Clamp |
| Material | Nickel-plated |
| Inkluderar | Shelf Clamp |
| Beskrivning | Clamp, Specialty, Flex, CLS-FLEXBM |
| Diameter (metrisk) | 13 mm |
| Längd (engelska) Arm | 12 in. |
| Längd (metrisk) arm | 305 mm |
| Storlek (metriskt) grepp | 0 to 43 mm |
| Typ av käke | 3-Prong Dual Adjust |