Learn More
JULABO™ Flexible Metal Tubing
For use with PRESTO™ Temperature control system/process system. Triple insulated for use up to 350°C.
5695.00 SEK - 30215.00 SEK
Specifications
Description | Metal tubing |
---|---|
Temperature Range | -100°C to +350°C |
Material | Metal |
Product Code | Brand | Type | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Size | Price | Quantity & Availability | ||||
15274313
|
JULABO™
8930262 |
Connection: 2 fittings M24 x 1.5 female | 1.5 m |
7245.00 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15264313
|
JULABO™
8930261 |
Connection: 2 fittings M24 x 1.5 female | 1 m |
5695.00 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15284313
|
JULABO™
8930263 |
Connection: 2 fittings M24 x 1.5 female | 2 m |
8630.00 SEK
Pack of 2 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15204323
|
JULABO™
8930271 |
Connection: 2 fittings M30 x 1.5 female | 1 m |
8655.00 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15234323
|
JULABO™
8930274 |
Connection: 2 fittings M30 x 1.5 female | 3 m |
16285.00 SEK
Pack of 3 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15214323
|
JULABO™
8930272 |
Connection: 2 fittings M30 x 1.5 female | 1.5 m |
10710.00 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15224323
|
JULABO™
8930273 |
Connection: 2 fittings M30 x 1.5 female | 2 m |
12545.00 SEK
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
15244323
|
JULABO™
8930275 |
Connection: 2 fittings M30 x 1.5 female | 5 m |
25350.00 SEK
Pack of 5 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15254323
|
JULABO™
8930282 |
Connection: 2 fittings M38 x 1.5 female | 1.5 m |
12450.00 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15264323
|
JULABO™
8930283 |
Connection: 2 fittings M38 x 1.5 female | 2 m |
16015.00 SEK
Pack of 2 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15274323
|
JULABO™
8930284 |
Connection: 2 fittings M38 x 1.5 female | 3 m |
19790.00 SEK
Pack of 3 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15284323
|
JULABO™
8930285 |
Connection: 2 fittings M38 x 1.5 female | 5 m |
30215.00 SEK
Pack of 5 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
Description
- Flexible Metal tubing
- Triple insulated
- -100 ... +350 °C.
Available lengths and fittings:
- 1 m:
- M24 x 1.5 female
- M30 x 1.5 female
- 1.5 m:
- M24 x 1.5 female
- M30 x 1.5 female
- M38 x 1.5 female
- 2 m:
- M24 x 1.5 female
- M30 x 1.5 female
- M38 x 1.5 female
- 3 m:
- M30 x 1.5 female
- M38 x 1.5 female
- 5 m:
- M30 x 1.5 female
- M38 x 1.5 female
Specifications
Metal tubing | |
-100°C to +350°C | |
Metal |