Learn More
Memmert™ Flexible Pt Temperature Sensor
For Use With: ICP750
Brand:  Memmert™ ICP750H8
Additional Details : Weight : 0.11000kg
Product Code. 15943730
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Flexible Pt100 temperature sensor, positioned flexibly in chamber or load, for local temperature measurement (up to 2 additional sensors are possible)
  • The measured temperature can, if required, be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
Specifications
Specifications
ICP750
Temperature Sensor