Läs mer
Flibanserin Hydrochloride, TRC
Produktkod. 30133880
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133880 25 mg
30134169 250 mg
30007128 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133880 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF40127025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 147359-76-0
Molekylformel C20 H21 F3 N4 O . Cl H
Molekylvikt (g/mol) 426.86
Synonym 2H-Benzimidazol-2-one, 1,3-dihydro-1-[2-[4-[3-(trifluoromethyl)phenyl]-1-piperazinyl]ethyl]-, hydrochloride (1:1), 2H-Benzimidazol-2-one, 1,3-dihydro-1-[2-[4-[3-(trifluoromethyl)phenyl]-1-piperazinyl]ethyl]-, monohydrochloride (9CI), Flibanserin hydrochloride
IUPAC-namn 3-[2-[4-[3-(trifluoromethyl)phenyl]piperazin-1-yl]ethyl]-1H-benzimidazol-2-one;hydrochloride
LEDER Cl.FC(F)(F)c1cccc(c1)N2CCN(CCN3C(=O)Nc4ccccc34)CC2
Specifikationer
Kemiskt namn eller material Flibanserin Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C20H21F3N4O.ClH/c21-20(22,23)15-4-3-5-16(14-15)26-11-8-25(9-12-26)10-13-27-18-7-2-1-6-17(18)24-19(27)28;/h1-7,14H,8-13H2,(H,24,28);1H
Formel vikt 426.1434