missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FLJ20399 Recombinant Protein
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH06527 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 54920 |
Molecular Weight (g/mol) | 79.97 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180893
|
Abnova™
H00054920-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190893
|
Abnova™
H00054920-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH06527 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
79.97 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DUS2L | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
54920 | |
FLJ20399 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MILNSLSLCYHNKLILAPMVRVGTLPMRLLALDYGADIVYCEELIDLKMIQCKRVVNEVLSTVDFVAPDDRVVFRTCEREQNRVVFQMGTSDAERALAVARLVENDVAGIDVNMGCPKQYSTKGGMGAALLSDPDKIEKILSTLVKGTRRPVTCKIRILPSLEDTLSLVKRIERTGIAAIAVHGRKREERPQHPVSCEVIKAIADTLSIPVIANGGSHDHIQQYSDIEDFRQATAASSVMVARAAMWNPSIFLKEGLRPLEEVMQKYIRYAVQYDNHYTNTKYCLCQMLREQLESPQGRLLHAAQSSREICEAFGLGAFYEETTQELDAQQARLSAKTSEQTGEPAEDTSGVIKMAVKFDRRAYPAQITPKMCLLEWCRREKLAQPVYETVQRPLDRLFSSIVTVAEQKYQSTLWDKSKKLAEQAAAIVCLRSQGLPEGRLGEESPSLHKRKREAPDQDPGGPRAQELAQPGDLCKKPFVALGSGEESPLEGW | |
DUS2/FLJ20399/SMM1/URLC8 | |
DUS2L | |
Wheat Germ (in vitro) | |
GST |