missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ25006 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26103-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
FLJ25006 likely exerts a protein serine/threonine kinase activity.Specifications
FLJ25006 | |
Polyclonal | |
Unconjugated | |
SGK494 | |
FLJ25006, SGK494 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124923, 216976 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5SYL1, Q96LW2 | |
Rskr, SGK494 | |
FLJ25006 Fusion Protein Ag22017 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |