Läs mer
Systec™ Floor Model, Pass-Through Autoclave, HX Series 2D
Produktkod. 15335905
Change view
Kapacitet (metrisk):
209 l (totalt), 200 l (nominellt)
370 l (totalt), 320 l (nominellt)
Djup (metrisk) kammare:
100 mm
750 mm
Diameter (metrisk) kammare:
500 mm
740 mm
Modell:
HX-200 2D
HX-320 2D
Förpackningsstorlek:
Varje
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kapacitet (metrisk) Djup (metrisk) kammare Diameter (metrisk) kammare Modell unitSize
15335905 209 l (totalt), 200 l (nominellt) 100 mm 500 mm HX-200 2D Varje
12572767 370 l (totalt), 320 l (nominellt) 750 mm 740 mm HX-320 2D Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15335905 Leverantör Systec™ Leverantörsnummer 2207
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Systec™ Floor Model, Pass-Through Autoclave, HX Series 2D are designed to be built into a wall for clean room operations.
  • Highest requirements with regard to safe, accurate, reproducible and validatable sterilization processes.
  • For use under the most stringent clean room and safety conditions: For biological safety laboratories, e.g., fitting as a sterilization and pass-through lock for protecting the external environment, for clean rooms in laboratories and production facilities as a sterilization and pass-through lock separating sterile and non-sterile areas.
  • Fully automatic, micro-processor controlled.
  • Can be used for all laboratory applications, even for sophisticated sterilization processes.
  • All additional optional accessories can be fitted to obtain validatable processes.
  • Standard features:
    • Integrated, separate steam generator
    • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
    • Touch-Screen control
    • Number of sterilization programs: up to 100
    • Code-secured access rights for changing parameters and further safety-relevant intervention
    • Internal memory for storing up to 500 sterilization cycles
    • Timer for starting programs
    • Autofill: automatic demineralized water feed for steam generation
    • Flexible PT-100 temperature sensor
    • Additional temperature sensor in condense exhaust
    • Temperature holding function for liquids after program finish
    • Special program for Durham tubes
    • Calculation of F0 value
    • Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
    • Water-cooled steam exhaust, thermostatically controlled
    • Programmable automatic door-opening on completion of program
    • RS-232 and RS-485 interfaces for external data transmission (network-compatible)
  • Available options include temperature, pressure, cooling, vacuum, drying, exhaust, and documentation options.
Specifikationer
Kapacitet (metrisk) 209 l (totalt), 200 l (nominellt)
Djup (metrisk) kammare 100 mm
Diameter (metrisk) kammare 500 mm
Modell HX-200 2D
Produkttyp Double-Door/Pass-Through Autoclave
Temperatur (metrisk) 140°C
Driftstryck 4 bar