missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Flotillin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528118
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Pig (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
Specifikationer
| Flotillin 1 | |
| Polyclonal | |
| Unconjugated | |
| FLOT1 | |
| DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100763504, 10211, 14251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.34 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O08917, O75955 | |
| FLOT1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Flotillin 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Hamster, Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering