missing translation for 'onlineSavingsMsg'
Learn More

Flotillin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15571-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824804

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes an protein that localizes to the caveolae, which are small domains on the inner cell membranes. This protein plays a role in vesicle trafficking and cell morphology. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

Flotillin 1
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08917, O75955, Q9Z1E1
FLOT1, flotillin 1
Rabbit
IgG
150 μL
-20°C
Primary
10211, 14251, 64665
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
FLOT1
Liquid
Flot1
Flotillin 1 Fusion Protein Ag7902
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers