Learn More
Thermo Scientific™ Temperature Flow and Pressure Control Accessories
Refine the pressure and flow characteristics of your Thermo Scientific™ recirculating chiller.
Brand: Thermo Scientific™ 611000000106
Additional Details : Weight : 0.01480kg
Description
Customize the flow and pressure output of recirculating chiller to match the needs of your process. Available in several popular sizes Thermo Scientific™ Flow and pressure control kits for Thermo Scientific chillers can have you up and running without the hassle of finding parts.
- Designed to work with your Thermo Scientific recirculating chiller
- Available in popular sizes that match your recirculating chiller connections and interface with your process
- Take the guess work out of finding parts to match your recirculating chiller connections
- Designed to match the operating range of your recirculating chiller
Additional questions? Please contact Thermo Scientific Technical support if you’re unsure how best to refine the flow and pressure of your recirculating chiller.
Specifications
External Flow Control Kit | |
ThermoFlex™ 900 - 10000 with P1, P2 or T1 Pumps | |
Flow Control Kit | |
1/2 in. NPT |