Läs mer
IBA Lifesciences Flow Restrictor
Produktkod. 17297590 Handla allt IBA Lifesciences Produkter
Kvantitet:
5/Pk.
Förpackningsstorlek:
Paket med 5
Produktkod. 17297590
Brand: IBA Lifesciences 66310999
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Flow restrictor for 1 ml Strep-Tactin™ TACS Agarose Column
  • The flow restrictors can be used during sample loading in our Fab-TACS™ technology for cells.
  • They fit on 1 mL Strep-Tactin™ TACS Agarose columns.
Specifikationer
Kvantitet 5/Pk.