missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLT3 [D835Y] Recombinant Human Protein, Invitrogen™
FLT3 [D835Y] Recombinant Human Protein
Brand: Invitrogen PV3967
Additional Details : Weight : 0.00945kg
Description
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family. The FLT3 D835Y mutant is a drug resistant form of FLT3.Specifications
NP_004110, D835Y | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
48.6kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
FLT3 D835Y, FLK2, STK1, CD135 | |
FLT3 D835Y was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 564-958; D835Y | |
7,160nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
His Tag | |
Kinase and inhibitors | |
65% by SDS-PAGE |
0.2mg/mL | |
2322, 4790 | |
FLT3 [D835Y] Recombinant Human Protein | |
–80°C | |
1 Each | |
FLT3 [D835Y] Recombinant Human Protein | |
PDGFR | |
FLT3 [D835Y] Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.