Läs mer
Fludarabine, TRC
Produktkod. 30136192
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136192 100 mg
30133679 25 mg
30011468 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136192 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF425000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21679-14-1
Molekylformel C10 H12 F N5 O4
Molekylvikt (g/mol) 285.23
Synonym 9H-Purin-6-amine, 9-β-D-arabinofuranosyl-2-fluoro-, Adenine, 9-β-D-arabinofuranosyl-2-fluoro- (8CI), 9-β-D-Arabinofuranosyl-2-fluoro-9H-purin-6-amine, 2-Fluoro Ara-A, 2-Fluoro-9-β-D-arabinofuranosyladenine, 2-Fluoroadenine arabinoside, 2F-Ara-A, 9-β-D-Arabinofuranosyl-2-fluoroadenine, 9-β-D-Arabinosyl-2-fluoroadenine, F-ara-A, Fludarabine, NSC 118218, NSC 118218H
IUPAC-namn (2R,3S,4S,5R)-2-(6-amino-2-fluoropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
LEDER Nc1nc(F)nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O
Specifikationer
Kemiskt namn eller material Fludarabine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C10H12FN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6+,9-/m1/s1
Formel vikt 285.0873
Procent renhet >95%