Läs mer
Fludrocortisone Acetate, Mikromol™
Produktkod. 18010030
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
18010030 250 mg
1 options
Produktkod. 18010030 Leverantör Mikromol Leverantörsnummer MM0817.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Fludrocortisone Acetate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fludrocortisone Acetate
CAS 514-36-3
Analyt- eller komponentnamn Fludrocortisone Acetate
Frakt skick Room Temperature
Molekylformel C23 H31 F O6
Certifikat och efterlevnad ISO 17034
Synonym Fludrocortisone acetate, Corticosterone, 9-fluoro-17-hydroxy-, 21-acetate (6CI), 9-Fluorohydrocortisone acetate, Alflorone acetate, Cortineff, Fludrocortisone 21-acetate, Fluorocortisol acetate, Fluorohydrocortisone acetate, NSC 15186, Scherofluron, U 4845, 9-Fluoro-11beta,17-dihydroxy-3,20-dioxopregn-4-en-21-yl acetate, Pregn-4-ene-3,20-dione, 9-fluoro-11beta,17,21-trihydroxy-, 21-acetate (7CI,8CI), 9alpha-Fluoro-11beta,17alpha,21-trihydroxypregn-4-ene-3,20-dione 21-acetate, 9alpha-Fluorocortisol 21-acetate, 9alpha-Fluorocortisol acetate, 9alpha-Fluorohydrocortisone 21-acetate, 9alpha-Fluorohydrocortisone acetate
InChI formel InChI=1S/C23H31FO6/c1-13(25)30-12-19(28)22(29)9-7-16-17-5-4-14-10-15(26)6-8-20(14,2)23(17,24)18(27)11-21(16,22)3/h10,16-18,27,29H,4-9,11-12H2,1-3H3/t16-,17-,18-,20-,21-,22-,23-/m0/s1
LEDER CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,17R)-9-fluoro-11,17-dihydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molekylvikt (g/mol) 422.49
Kvantitet 250 mg
Formel vikt 422.2105 g/mol