Läs mer
Fludrocortisone Acetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Fludrocortisone Acetate |
| CAS | 514-36-3 |
| Molekylformel | C23 H31 F O6 |
| Kvantitet | 10 g |
| Synonym | Fludrocortisone acetate, 9-Fluoro-11β,17-dihydroxy-3,20-dioxopregn-4-en-21-yl acetate, Corticosterone, 9-fluoro-17-hydroxy-, 21-acetate (6CI), Pregn-4-ene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-, 21-acetate (7CI,8CI), 9-Fluorohydrocortisone acetate, 9α-Fluoro-11β,17α,21-trihydroxypregn-4-ene-3,20-dione 21-acetate, 9α-Fluorocortisol 21-acetate, 9α-Fluorocortisol acetate, 9α-Fluorohydrocortisone 21-acetate, 9α-Fluorohydrocortisone acetate, Alflorone acetate, Cortineff, Fludrocortisone 21-acetate, Fludrocortisone acetate, Fluorocortisol acetate, Fluorohydrocortisone acetate, NSC 15186, Scherofluron, U 4845 |
| InChI formel | InChI=1S/C23H31FO6/c1-13(25)30-12-19(28)22(29)9-7-16-17-5-4-14-10-15(26)6-8-20(14,2)23(17,24)18(27)11-21(16,22)3/h10,16-18,27,29H,4-9,11-12H2,1-3H3/t16-,17-,18-,20-,21-,22-,23-/m0/s1 |
| LEDER | CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C |
| IUPAC-namn | [2-[(8S,9R,10S,11S,13S,14S,17R)-9-fluoro-11,17-dihydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Molekylvikt (g/mol) | 422.49 |
| Formel vikt | 422.2105 |