Läs mer
Fludrocortisone, TRC
Produktkod. 17981825
Change view
Kvantitet:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981825 10 mg
17912375 50 mg
17910585 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17981825 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF42810010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127-31-1
Molekylformel C21 H29 F O5
Molekylvikt (g/mol) 380.45
Synonym Pregn-4-ene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy- (7CI,8CI), 9-Fluoro-11β,17,21-trihydroxypregn-4-ene-3,20-dione, 9-Fluorocortisol, 9-Fluorohydrocortisone, 9α-Fludrocortisone, 9α-Fluorcortisol, 9α-Fluoro-11β,17α,21-trihydroxy-4-pregnene-3,20-dione, 9α-Fluoro-17-hydroxycorticosterone, 9α-Fluorocortisol, 9α-Fluorohydrocortisone, Alflorone, Astonin-H, F-COL, F-Cortef, Florinef, Fludrocortisone, Fludrocortone, Fludrone, Fludronef, Fluodrocortisone, Fluohydrisone, Fluohydrocortisone, Fluorocortisol, Fluorocortisone, Fluorohydrocortisone, NSC 11318, Pregn-4-ene-3,20-dione, 9-fluoro-11,17,21-trihydroxy-, (11β)-
IUPAC-namn (8S,9R,10S,11S,13S,14S,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Specifikationer
Kemiskt namn eller material Fludrocortisone
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C21H29FO5/c1-18-7-5-13(24)9-12(18)3-4-15-14-6-8-20(27,17(26)11-23)19(14,2)10-16(25)21(15,18)22/h9,14-16,23,25,27H,3-8,10-11H2,1-2H3/t14-,15-,16-,18-,19-,20-,21-/m0/s1
Formel vikt 380.1999
Procent renhet >95%