Läs mer
Flufenacet ESA Sodium Salt, TRC
Produktkod. 30406093
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30406093 25 mg
1 options
Produktkod. 30406093 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF59942825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Flufenacet ethane sulfonic acid ESA sodium salt
CAS 947601-87-8
Molekylformel C11 H13 F N O4 S . Na
Kvantitet 25 mg
Synonym Flufenacet-ethane sulfonic acid (ESA) sodium, Ethanesulfonic acid, 2-[(4-fluorophenyl)(1-methylethyl)amino]-2-oxo-, sodium salt (1:1)
InChI formel InChI=1S/C11H14FNO4S.Na/c1-8(2)13(11(14)7-18(15,16)17)10-5-3-9(12)4-6-10;/h3-6,8H,7H2,1-2H3,(H,15,16,17);/q;+1/p-1
LEDER [Na+].CC(C)N(C(=O)CS(=O)(=O)[O-])c1ccc(F)cc1
IUPAC-namn sodium;2-(4-fluoro-N-propan-2-ylanilino)-2-oxoethanesulfonate
Molekylvikt (g/mol) 297.28
Formel vikt 297.0447