Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Flufenacet |
| Förpackning | Glass Bottle |
| CAS | 142459-58-3 |
| Färg | Beige |
| Smältpunkt | 76°C to 77°C |
| Kokpunkt | Decomposes on heating. |
| Synonym | N-(4-fluorophenyl)-N-(1-methylethyl)-2-[[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]oxy]acetamide |
| Industry Type | Environmental Analysis |
| Molekylvikt (g/mol) | 363.33 |
| Kvantitet | 100 mg |
| Visa mer |