Läs mer
Flumatinib, MedChemExpress
Produktkod. 16498537
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16498537 5 mg -
16415908 10 mg 10mg
16402057 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16498537 Leverantör Medchem Express Leverantörsnummer HY13904
MedChemExpress Flumatinib (HHGV678) is an orally available, selective inhibitor of Bcr-Abl. Flumatinib inhibits c-Abl, PDGFRβ and c-Kit with IC50s of 1.2 nM, 307.6 nM and 665.5 nM, respectively.
Flumatinib (HHGV678) is an orally available, selective inhibitor of Bcr-Abl. Flumatinib inhibits c-Abl, PDGFRβ and c-Kit with IC50s of 1.2 nM, 307.6 nM and 665.5 nM, respectively.
Specifikationer
Kemiskt namn eller material Flumatinib
CAS 895519-90-1
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C29H29F3N8O
Kvantitet 5 mg
Synonym HHGV678
Löslighetsinformation DMSO : 100 mg/mL (177.75 mM; Need ultrasonic)
LEDER O=C(NC1=CC(NC2=NC=CC(C3=CC=CN=C3)=N2)=C(C)N=C1)C4=CC=C(CN5CCN(C)CC5)C(C(F)(F)F)=C4
Molekylvikt (g/mol) 562.59
Formel vikt 562.59
Procent renhet 98.1%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-Kinase/protease