Läs mer
Flumequine EP Impurity B, Mikromol™
Produktkod. 30050408
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050408 25 mg
30051601 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050408 Leverantör Mikromol Leverantörsnummer MM0842.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Flumequine EP Impurity B
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Flumequine Ethyl Ester
CAS 42835-47-2
Analyt- eller komponentnamn Flumequine Ethyl Ester
Frakt skick Room Temperature
Molekylformel C16 H16 F N O3
Certifikat och efterlevnad ISO 9001
Synonym Ethyl (RS)-9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-benzo[i,j]quinolizine-2-carboxylate (Flumequine Ethyl Ester), Flumequine Imp. B (EP), 9-fluoro-6,7-dihydro-5-methyl-1-oxo-1H,5H-Benzo[ij]quinolizine-2-carboxylic acid ethyl ester, Flumequine ethyl ester, Flumequine Impurity B
InChI formel InChI=1S/C16H16FNO3/c1-3-21-16(20)13-8-18-9(2)4-5-10-6-11(17)7-12(14(10)18)15(13)19/h6-9H,3-5H2,1-2H3
LEDER CCOC(=O)C1=CN2C(C)CCc3cc(F)cc(C1=O)c23
IUPAC-namn Ethyl (RS)-9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-benzo[i,j]quinolizine-2-carboxylate
Molekylvikt (g/mol) 289.3
Kvantitet 25 mg
Formel vikt 289.1114 g/mol