Läs mer
Flumethasone 21-Pivalate, TRC
Produktkod. 17962375
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17962375 100 mg
17903515 25 mg
17921505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17962375 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF455015100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2002-29-1
Molekylformel C27 H36 F2 O6
Molekylvikt (g/mol) 494.57
Synonym 6α,9-Difluoro-11β,17-dihydroxy-16α-methyl-3,20-dioxopregna-1,4-dien-21-yl 2,2-dimethylpropanoate, Pregna-1,4-diene-3,20-dione, 21-(2,2-dimethyl-1-oxopropoxy)-6,9-difluoro-11,17-dihydroxy-16-methyl-, (6α,11β,16α)-, Pregna-1,4-diene-3,20-dione, 6α,9-difluoro-11β,17,21-trihydroxy-16α-methyl-, 21-pivalate (7CI,8CI), Pivalic acid, 21-ester with 6α,9-difluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione (8CI), 6α,9α-Difluoro-11β,17α,21-trihydroxy-16α-methyl-1,4-pregnadiene-3,20-dione 21-pivalate, 6α,9α-Difluoro-16α-methylprednisolone 21-pivalate, Flumetasone 21-pivalate, Flumetasone pivalate, Flumethasone 21-pivalate, Flumethasone pivalate, Locacorten, Locorten, Lorinden, Losalen, NSC 107680
IUPAC-namn [2-[(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2-dimethylpropanoate
LEDER C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C(C)(C)C
Specifikationer
Kemiskt namn eller material Flumetasone pivalate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C27H36F2O6/c1-14-9-16-17-11-19(28)18-10-15(30)7-8-24(18,5)26(17,29)20(31)12-25(16,6)27(14,34)21(32)13-35-22(33)23(2,3)4/h7-8,10,14,16-17,19-20,31,34H,9,11-13H2,1-6H3/t14-,16+,17+,19+,20+,24+,25+,26+,27+/m1/s1
Formel vikt 494.248
Procent renhet >95%