Läs mer
Flumethasone, TRC
Produktkod. 18091420
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18091420 50 mg
18070820 250 mg
18082210 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18091420 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF45500050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2135-17-3
Molekylformel C22 H28 F2 O5
Molekylvikt (g/mol) 410.45
Synonym Flumetasone, Flumetasone Pivalate Imp. A (EP), Pregna-1,4-diene-3,20-dione, 6,9-difluoro-11,17,21-trihydroxy-16-methyl-, (6α,11β,16α)-, Pregna-1,4-diene-3,20-dione, 6α,9-difluoro-11β,17,21-trihydroxy-16α-methyl- (8CI), (6α,11β,16α)-6,9-Difluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 6α,9-Difluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione, 6α,9α-Difluoro-16α-methyl-1,4-pregnadiene-11β,17α,21-triol-3,20-dione, 6α,9α-Difluoro-16α-methylprednisolone, 6α-Fluorodexamethasone, 6α-Fluorodexamethazone, Anaprime, Aniprime, Aniprome, Cortexillar, Flucort, Flucorticin, Flumethason, Flumethasone, Flutamethasone, Fluvet, Methagon, NSC 5402, NSC 54702, RS 2177, U 10974, Vecort
IUPAC-namn (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone)
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C22H28F2O5/c1-11-6-13-14-8-16(23)15-7-12(26)4-5-19(15,2)21(14,24)17(27)9-20(13,3)22(11,29)18(28)10-25/h4-5,7,11,13-14,16-17,25,27,29H,6,8-10H2,1-3H3/t11-,13+,14+,16+,17+,19+,20+,21+,22+/m1/s1
Formel vikt 410.1905
Procent renhet >95%