Läs mer
Fluocinolone Acetonide-21-aldehyde, TRC
Produktkod. 30009215
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009215 50 mg
30010874 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF45581050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13242-30-3
Molekylformel C24 H28 F2 O6
Molekylvikt (g/mol) 450.47
Synonym Pregna-1,4-dien-21-al, 6,9-difluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-3,20-dioxo-, (6α,11β,16α)-, Pregna-1,4-dien-21-al, 6α,9-difluoro-11β,16α,17-trihydroxy-3,20-dioxo-, cyclic 16,17-acetal with acetone (7CI,8CI), (6α,11β,16α)-6,9-Difluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-3,20-dioxopregna-1,4-dien-21-al, 6α,9α-Difluoro-11β,16α,17α-trihydroxypregna-1,4-diene-3,20-dione-21-al-16,17-acetonide, Fluocinolone acetonide-21-aldehyde, 6α,9-Difluoro-11β-hydroxy-16α,17-(1-methylethylidenedioxy)-3,20-dioxopregna-1,4-dien-21-al
LEDER CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)C=O
Specifikationer
Kemiskt namn eller material Fluocinolone acetonide-21-aldehyde
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C24H28F2O6/c1-20(2)31-19-9-13-14-8-16(25)15-7-12(28)5-6-21(15,3)23(14,26)17(29)10-22(13,4)24(19,32-20)18(30)11-27/h5-7,11,13-14,16-17,19,29H,8-10H2,1-4H3/t13-,14-,16-,17-,19+,21-,22-,23-,24+/m0/s1
Formel vikt 450.1854
Procent renhet >95%