Läs mer
Fluocinolone (Acetonide), MedChemExpress
Produktkod. 16492159
Encompass_Preferred
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16492159 1 g
16470918 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16492159 Leverantör Medchem Express Leverantörsnummer HYB0415
MedChemExpress Fluocinolone Acetonide is a glucocorticoid derivative used topically in the treatment of various skin disorders.
Fluocinolone Acetonide is a glucocorticoid derivative used topically in the treatment of various skin disorders.
Specifikationer
Kemiskt namn eller material Fluocinolone (Acetonide)
CAS 67-73-2
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C24H30F2O6
Kvantitet 1 g
Löslighetsinformation DMSO : ≥ 31 mg/mL (68.51 mM)
LEDER O=C([C@]([C@@]1([H])C[C@@]2([H])[C@@](C[C@H](F)C3=CC4=O)([H])[C@@](F)([C@]3(C=C4)C)[C@@H](O)C5)(OC(C)(C)O1)[C@]25C)CO
Molekylvikt (g/mol) 452.49
Formel vikt 452.49
Procent renhet 97.98%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation