Läs mer
Fluocinonide 22 Methyl Homologue, TRC
Produktkod. 30405329
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30405329 50 mg
1 options
Produktkod. 30405329 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF45582650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fluocinonide 22 Methyl Homologue
CAS 51333-59-6
Molekylformel C25H30F2O7
Kvantitet 50 mg
Synonym 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione derivative
InChI formel InChI=1S/C25H30F2O7/c1-12(28)32-11-20(31)25-21(33-13(2)34-25)9-15-16-8-18(26)17-7-14(29)5-6-22(17,3)24(16,27)19(30)10-23(15,25)4/h5-7,13,15-16,18-19,21,30H,8-11H2,1-4H3/t13?,15-,16-,18-,19-,21+,22-,23-,24-,25+/m0/s1
LEDER CC1O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)COC(=O)C
Molekylvikt (g/mol) 480.5
Formel vikt 480.196