missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Fluorescein disodium salt hydrate
343.98 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.
Solubility
Soluble in water(500g/L).
Notes
Store in cool, dry conditions in well sealed containers.
Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.
Solubility
Soluble in water(500g/L).
Notes
Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | >250°C |
pH | 8.3 |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 3833041 |
Merck Index | 14,4159 |
Solubility Information | Soluble in water(500g/L). |
Formula Weight | 376.28 |
Chemical Name or Material | Fluorescein disodium salt hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'