Läs mer
Fluoresceinamine Maleic Acid Monoamide, TRC
Produktkod. 30407903
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30407903 100 mg
1 options
Produktkod. 30407903 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF485335100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fluoresceinamine Maleic Acid Monoamide
CAS 75900-74-2
Molekylformel C24 H15 N O8
Kvantitet 100 mg
Synonym (2Z)-4-[(3',6'-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthen]-5-yl)amino]-4-oxo-2-butenoic Acid
InChI formel InChI=1S/C24H15NO8/c26-13-2-5-17-19(10-13)32-20-11-14(27)3-6-18(20)24(17)16-4-1-12(9-15(16)23(31)33-24)25-21(28)7-8-22(29)30/h1-11,26-27H,(H,25,28)(H,29,30)/b8-7-
LEDER OC(=O)\C=C/C(=O)Nc1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35
IUPAC-namn (Z)-4-[(3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-yl)amino]-4-oxobut-2-enoic acid
Molekylvikt (g/mol) 445.378
Formel vikt 445.08