Läs mer
Fluorescent Brightener 220, Technical Grade, TRC
Produktkod. 30405880
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30405880 500 mg
1 options
Produktkod. 30405880 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF597828500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fluorescent Brightener 220
CAS 16470-24-9
Molekylformel C40H40N12Na4O16S4
Kvantitet 500 mg
Synonym Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-[bis(2-hydroxyethyl)amino]-6-[(4-sulfophenyl)amino]-1,3,5-triazin-2-yl]amino]-, sodium salt (1:4), 2,2’-Stilbenedisulfonic acid, 4,4'-bis[[4-[bis(2-hydroxyethyl)amino]-6-(p-sulfoanilino)-s-triazin-2-yl]amino]-, tetrasodium salt (7CI,8CI), Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-[bis(2-hydroxyethyl)amino]-6-[(4-sulfophenyl)amino]-1,3,5-triazin-2-yl]amino]-, tetrasodium salt (9CI), BLB, Belofor OB, Blancofor BBU, Blankophor BBU, Blankophor P/BBU, Blankophor Z-JPN, C.I. 40623, C.I. Fluorescent Brightener 220, Kahablanc APU, Keyfluor White CN, Leucophor U, Photine CPK, SCRI Renaissance 2200, Sandoz CD, Tetrasodium 4,4'-bis[2-di(β-hydroxyethyl)amino-4-(4-sulfophenylamino)-s-triazin-6-ylamino]stilbine-2,2'-disulfonate
InChI formel InChI=1S/C40H44N12O16S4.4Na/c53-19-15-51(16-20-54)39-47-35(41-27-7-11-31(12-8-27)69(57,58)59)45-37(49-39)43-29-5-3-25(33(23-29)71(63,64)65)1-2-26-4-6-30(24-34(26)72(66,67)68)44-38-46-36(48-40(50-38)52(17-21-55)18-22-56)42-28-9-13-32(14-10-28)70(60,61)62;;;;/h1-14,23-24,53-56H,15-22H2,(H,57,58,59)(H,60,61,62)(H,63,64,65)(H,66,67,68)(H2,41,43,45,47,49)(H2,42,44,46,48,50);;;;/q;4*+1/p-4
LEDER OCCN(C1=NC(NC2=CC=C(S(=O)(O[Na])=O)C=C2)=NC(NC3=C(S(=O)(O[Na])=O)C=C(/C=C/C4=CC(S(=O)(O[Na])=O)=C(NC5=NC(NC6=CC=C(S(=O)(O[Na])=O)C=C6)=NC(N(CCO)CCO)=N5)C=C4)C=C3)=N1)CCO
IUPAC-namn tetrasodium;5-[[4-[bis(2-hydroxyethyl)amino]-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-[2-[4-[[4-[bis(2-hydroxyethyl)amino]-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]ethenyl]benzenesulfonate
Molekylvikt (g/mol) 1165.04
Formel vikt 1164.1159