Läs mer
Fluoride Ion Chromatography Standard: F- @ 100μg/mL in H2O, VHG™
Produktkod. 17978874
Change view
Kvantitet:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17978874 500 ml
17949914 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17978874 Leverantör VHG Labs Leverantörsnummer VHGIF100500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Fluoride Ion Chromatography Standard: F- @ 100μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fluoride
CAS 16984-48-8
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnamn Fluoride
Frakt skick Room Temperature
Molekylformel F-
Certifikat och efterlevnad ISO 17034
Synonym F−
LEDER [F-]
IUPAC-namn Fluoride
Lösningsmedel eller matris Water
Molekylvikt (g/mol) 19
Kvantitet 500 ml
Formel vikt 19 g/mol