Läs mer
Fluorol Yellow 088, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Fluorol Yellow 088 |
| CAS | 81-37-8 |
| Molekylformel | C22H16O |
| Kvantitet | 500 mg |
| Synonym | Fluorol 5G, 2,8-Dimethylnaphtho[3,2,1-kl]xanthene, 2,8-Dimethylceroxene, C.I. 45550, C.I. Fluorescent Brightener 74, C.I. Fluorescent Brightening Agent 74, C.I. Solvent Green 4, Fluorescent Brightener 74, Fluorescent Green Dye, Fluorescent Green LF, Solvent Green 4 |
| InChI formel | InChI=1S/C22H16O/c1-13-7-9-19-18(11-13)21-16-6-4-3-5-15(16)12-17-14(2)8-10-20(23-19)22(17)21/h3-12H,1-2H3 |
| LEDER | CN([C@@H]1[C@@H](N2CCCC2)C[C@]3(CCCO3)CC1)C(CC4=CC(Cl)=C(Cl)C=C4)=O |
| Molekylvikt (g/mol) | 296.36 |
| Formel vikt | 296.1201 |