Läs mer
Fluorometholone, TRC
Produktkod. 30134131
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134131 500 mg
17904435 100 mg
17912535 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134131 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF593145500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 426-13-1
Molekylformel C22 H29 F O4
Molekylvikt (g/mol) 376.46
Synonym 9-Fluoro-11beta,17-dihydroxy-6alpha-methylpregna-1,4-diene-3,20-dione, 21-Desoxy-9alpha-fluoro-6-methylprednisolone, 6alpha-Methyl-9alpha-fluoro-11beta,17alpha-dihydroxypregna-1,4-diene-3,20-dione, 6alpha-Methyl-9alpha-fluoro-21-desoxyprednisolone, 9alpha-Fluoro-11beta,17alpha-dihydroxy-6alpha-methylpregna-1,4-diene-3,20-dione, Cortilet, Delmeson, Efflumidex, FML, Fluaton, Fluaton PVA, Flumetholon, Flumetholone, Fluor-Op, Fluormetholon, Fluorometholone, Loticort, NSC-33001, Oxylone, U 8614, Ursnon
IUPAC-namn (6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11,17-dihydroxy-6,10,13-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)C)[C@@]3(C)C[C@H](O)[C@]2(F)[C@@]4(C)C=CC(=O)C=C14
Specifikationer
Kemiskt namn eller material Fluorometholone
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C22H29FO4/c1-12-9-17-15-6-8-21(27,13(2)24)20(15,4)11-18(26)22(17,23)19(3)7-5-14(25)10-16(12)19/h5,7,10,12,15,17-18,26-27H,6,8-9,11H2,1-4H3/t12-,15-,17-,18-,19-,20-,21-,22-/m0/s1
Formel vikt 376.205
Procent renhet >95%