Läs mer
Fluorosalan, TRC
Produktkod. 30404785
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30404785 100 mg
1 options
Produktkod. 30404785 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF595065100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fluorosalan
CAS 4776-06-1
Molekylformel C14H8Br2F3NO2
Kvantitet 100 mg
Synonym 3,5-Dibromo-2-hydroxy-N-[3-(trifluoromethyl)phenyl]-benzamide, 3,5-Dibromo-α,α,α-trifluoro-m-salicylotoluidide, 3,5-Dibromo-2-hydroxy-N-[3-(trifluoromethyl)phenyl]benzamide, 3,5-Dibromo-3'-(trifluoromethyl)salicylanilide, 3,5-Dibromo-α,α,α-trifluoro-m-salicylotoluidide, Fluorophene, Fluorsalan, NSC 166520, Vancide FP, WR 78550
InChI formel InChI=1S/C14H8Br2F3NO2/c15-8-5-10(12(21)11(16)6-8)13(22)20-9-3-1-2-7(4-9)14(17,18)19/h1-6,21H,(H,20,22)
LEDER Oc1c(Br)cc(Br)cc1C(=O)Nc2cccc(c2)C(F)(F)F
Molekylvikt (g/mol) 439.02