Läs mer
Fluphenazine Decanoate S-oxide, TRC
Produktkod. 30406023
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30406023 25 mg
1 options
Produktkod. 30406023 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF59952525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fluphenazine Decanoate S-oxide
CAS 64610-50-0
Molekylformel C32H44F3N3O3S
Kvantitet 25 mg
Synonym 2-(4-(3-(5-Oxido-2-(trifluoromethyl)-10H-phenothiazin-10-yl)propyl)piperazin-1-yl)ethyl Decanoate
InChI formel InChI=1S/C32H44F3N3O3S/c1-2-3-4-5-6-7-8-14-31(39)41-24-23-37-21-19-36(20-22-37)17-11-18-38-27-12-9-10-13-29(27)42(40)30-16-15-26(25-28(30)38)32(33,34)35/h9-10,12-13,15-16,25H,2-8,11,14,17-24H2,1H3
LEDER FC(F)(F)C1=CC=C2S(C3=CC=CC=C3N(CCCN4CCN(CCOC(CCCCCCCCC)=O)CC4)C2=C1)=O
Molekylvikt (g/mol) 607.77
Formel vikt 607.3055