Läs mer
Fluphenazine N,N',S-Trioxide Dihydrochloride, Mikromol
Produktkod. 30460422
Change view
Kvantitet:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460422 4 x 25 mg
18072190 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30460422 Leverantör Mikromol Leverantörsnummer MM0114.0400254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Fluphenazine N,N',S-Trioxide Dihydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fluphenazine N,N',S-Trioxide Dihydrochloride
CAS 2732347-04-3
Analyt- eller komponentnamn Fluphenazine N,N',S-Trioxide Dihydrochloride
Frakt skick Room Temperature
Molekylformel C22 H26 F3 N3 O4 S . 2 Cl H
Certifikat och efterlevnad ISO 9001
Synonym Fluphenazine N,N',S-Trioxide Dihydrochloride
InChI formel InChI=1S/C22H26F3N3O4S.2ClH/c23-22(24,25)17-6-7-21-19(16-17)26(18-4-1-2-5-20(18)33(21)32)8-3-9-27(30)10-12-28(31,13-11-27)14-15-29;;/h1-2,4-7,16,29H,3,8-15H2;2*1H
Lösningstyp Neat
LEDER Cl.Cl.OCC[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3S(=O)c4ccc(cc24)C(F)(F)F)CC1
IUPAC-namn 2-[1,4-dioxido-4-[3-[5-oxo-2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazine-1,4-diium-1-yl]ethanol;dihydrochloride
Molekylvikt (g/mol) 558.44
Kvantitet 4 x 25 mg
Formel vikt 557.113 g/mol