Läs mer
Flupyrsulfuron-methyl, TRC
Produktkod. 30408418
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30408418 5 mg
1 options
Produktkod. 30408418 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5889305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Flupyrsulfuron-methyl
CAS 144740-53-4
Molekylformel C15 H14 F3 N5 O7 S
Kvantitet 5 mg
Synonym Flupyrsulfuron-methyl, 3-Pyridinecarboxylic acid, 2-[[[[(4,6-dimethoxy-2-pyrimidinyl)amino]carbonyl]amino]sulfonyl]-6-(trifluoromethyl)-, methyl ester, 2-[[[[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl]amino]sulfonyl]-6-(trifluoromethyl)-3-pyridinecarboxylic acid methyl ester
InChI formel InChI=1S/C15H14F3N5O7S/c1-28-9-6-10(29-2)21-13(20-9)22-14(25)23-31(26,27)11-7(12(24)30-3)4-5-8(19-11)15(16,17)18/h4-6H,1-3H3,(H2,20,21,22,23,25)
LEDER COC(=O)c1ccc(nc1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2)C(F)(F)F
IUPAC-namn methyl 2-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoyl]-6-(trifluoromethyl)pyridine-3-carboxylate
Molekylvikt (g/mol) 465.361
Formel vikt 465.057