Läs mer
Flurandrenolide, TRC
Produktkod. 30403118
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30403118 1 mg
1 options
Produktkod. 30403118 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5986501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Flurandrenolide
CAS 1524-88-5
Molekylformel C24 H33 F O6
Kvantitet 1 mg
Synonym Pregn-4-ene-3,20-dione, 6α-fluoro-11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (7CI,8CI), (6α,11β,16α)-6-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregn-4-ene-3,20-dione, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv., 33379, 6α-Fluoro-11β,21-dihydroxy-16α,17α-(isopropylidenedioxy)pregn-4-ene-3,20-dione, 6α-Fluoro-16α-hydroxyhydrocortisone 16,17-acetonide, Alondra F, Cordran, Drenison, Drocort, Fluadrenolone, Fludroxicortide, Fludroxycortide, Fluorandrenolone, Fluorandrenolone acetonide, Fluoroandrenolone acetonide, Flurandrenolide, Flurandrenolone, Flurandrenolone acetonide, Haelan, Haldrone F, L 33379, Sermaka
InChI formel InChI=1S/C24H33FO6/c1-21(2)30-19-9-14-13-8-16(25)15-7-12(27)5-6-22(15,3)20(13)17(28)10-23(14,4)24(19,31-21)18(29)11-26/h7,13-14,16-17,19-20,26,28H,5-6,8-11H2,1-4H3/t13-,14-,16-,17-,19+,20+,22-,23-,24+/m0/s1
LEDER CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)CC[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
IUPAC-namn (6α,11β,16α)-6-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregn-4-ene-3,20-dione
Molekylvikt (g/mol) 436.51
Formel vikt 436.2261