Läs mer
Flurofamide, TRC
Produktkod. 30240753
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240753 100 mg
30240967 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240753 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF596685100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70788-28-2
Molekylformel C7H9FN3O2P
Molekylvikt (g/mol) 217.14
Synonym 4-Fluoro-N-(diaminophosphinyl)benzamide, Flurfamide, NSC 342995
LEDER NP(=O)(N)NC(=O)c1ccc(F)cc1
Specifikationer
Kemiskt namn eller material Flurofamide
Kvantitet 100 mg
InChI formel InChI=1S/C7H9FN3O2P/c8-6-3-1-5(2-4-6)7(12)11-14(9,10)13/h1-4H,(H5,9,10,11,12,13)
Formel vikt 217.0416