Läs mer
Fluticasone, TRC
Produktkod. 30136803
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136803 5 mg
30006216 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136803 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5994565MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90566-53-3
Molekylformel C22 H27 F3 O4 S
Molekylvikt (g/mol) 444.51
Synonym Androsta-1,4-diene-17-carbothioic acid, 6,9-difluoro-11,17-dihydroxy-16-methyl-3-oxo-, S-(fluoromethyl) ester, (6α,11β,16α,17α)-, Fluticasone, VR 506, CAF (GW694301X), Fluticasone (GR51608X), CCI 22341, GW 872664X, TAF (GW675713X), 6-des-fluoro 698 (GW836850X), GSK465433A and GSK1322631A, 5-chloro-furoyl 698 (GSK180878A), TA-TAF dimer (GW871519X)
IUPAC-namn S-(fluoromethyl) (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carbothioate
LEDER C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)SCF
Specifikationer
Kemiskt namn eller material Fluticasone
Kvantitet 5 mg
InChI formel InChI=1S/C22H27F3O4S/c1-11-6-13-14-8-16(24)15-7-12(26)4-5-19(15,2)21(14,25)17(27)9-20(13,3)22(11,29)18(28)30-10-23/h4-5,7,11,13-14,16-17,27,29H,6,8-10H2,1-3H3/t11-,13+,14+,16+,17+,19+,20+,21+,22+/m1/s1
Formel vikt 444.1582