Läs mer
Flutrimazole, TRC
Produktkod. 30407584
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407584 500 mg
30406326 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30407584 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF622000500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Flutrimazole
CAS 119006-77-8
Molekylformel C22 H16 F2 N2
Kvantitet 500 mg
Synonym 1H-Imidazole, 1-[(2-fluorophenyl)(4-fluorophenyl)phenylmethyl]-, 1-[(2-Fluorophenyl)(4-fluorophenyl)phenylmethyl]-1H-imidazole, Flutrimazole, UR 4056
InChI formel InChI=1S/C22H16F2N2/c23-19-12-10-18(11-13-19)22(26-15-14-25-16-26,17-6-2-1-3-7-17)20-8-4-5-9-21(20)24/h1-16H
LEDER Fc1ccc(cc1)C(c2ccccc2)(c3ccccc3F)n4ccnc4
IUPAC-namn 1-[(2-fluorophenyl)-(4-fluorophenyl)-phenylmethyl]imidazole
Molekylvikt (g/mol) 346.37
Formel vikt 346.1282