Läs mer
FMK 9a, MedChemExpress
Produktkod. 16492596
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16492596 25 mg
16455797 5 mg
16449626 50 mg
16440669 1 mg
16414366 100 mg
16403068 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16492596 Leverantör Medchem Express Leverantörsnummer HY100522
MedChemExpress FMK 9a is an autophagin-1 inhibitor with IC50 values of 80 and 73 μM in FRET and LRA assay.
FMK 9a is an autophagin-1 inhibitor with IC50 values of 80 and 73 μM in FRET and LRA assay.
Specifikationer
Kemiskt namn eller material FMK 9a
Fysisk form Solid
CAS 1955550-51-2
Färg Off-White
Kvantitet 25 mg
Anteckningar om renhetsgrad Research
Molekylformel C23H21FN2O3
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 100 mg/mL (254.83 mM; Need ultrasonic)
LEDER O=C(C1=C(C=CC=C2)C2=CC=C1)N[C@H](C(NCC(CF)=O)=O)CC3=CC=CC=C3
Molekylvikt (g/mol) 392.42
Formel vikt 392.42
Procent renhet 98.0%