missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMNL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27834-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
27834-1-AP is a rabbit polyclonal antibody recognizes the 125-150 kDa protein.
NUCB2 was initially identified as calcium-binding protein EF hand motif-containing protein that could bind with Necdin, a growth suppressor expressed primarily in postmitotic neurons. NUC2 is a secreted protein that is cleaved into three major peptide products, Nesfatin-1, Nesfatin-2, and Nesfatin-3, of which Nesfatin-1 has been found to cause the suppression of food intake in a leptin-independent manner. Other studies have suggested that NUCB2/Nesfatin-1 may also play roles in energy homeostasis and closely related neuroendocrine functions.Specifications
FMNL1 | |
Polyclonal | |
Unconjugated | |
FMNL1 | |
C17orf1, C17orf1B, CLL associated antigen KW 13, FHOD4, FMNL, FMNL1, formin like 1, Formin like protein 1, KW 13, Leukocyte formin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
752 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95466 | |
FMNL1 | |
FMNL1 Fusion Protein Ag27312 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |