Läs mer
Fmoc-6-chloro L-Tryptophan, TRC
Produktkod. 30179010
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179010 10 mg
30178947 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179010 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF62800510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 908847-42-7
Molekylformel C26H21ClN2O4
Molekylvikt (g/mol) 460.91
Synonym 6-Chloro-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-tryptophan, Fmoc-6-chloro (S)-Tryptophan;
LEDER OC(=O)[C@H](Cc1c[nH]c2cc(Cl)ccc12)NC(=O)OCC3c4ccccc4c5ccccc35
Specifikationer
Kemiskt namn eller material Fmoc-6-chloro L-Tryptophan
InChI formel InChI=1S/C26H21ClN2O4/c27-16-9-10-17-15(13-28-23(17)12-16)11-24(25(30)31)29-26(32)33-14-22-20-7-3-1-5-18(20)19-6-2-4-8-21(19)22/h1-10,12-13,22,24,28H,11,14H2,(H,29,32)(H,30,31)/t24-/m0/s1
Kvantitet 10 mg
Formel vikt 460.119