Läs mer
Fmoc-a-methyl-L-phenylalanine, TRC
Produktkod. 30403165
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30403165 250 mg
1 options
Produktkod. 30403165 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF603535250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fmoc-alpha-methyl-L-phenylalanine
CAS 135944-05-7
Molekylformel C25H23NO4
Kvantitet 250 mg
InChI formel InChI=1S/C25H23NO4/c1-25(23(27)28,15-17-9-3-2-4-10-17)26-24(29)30-16-22-20-13-7-5-11-18(20)19-12-6-8-14-21(19)22/h2-14,22H,15-16H2,1H3,(H,26,29)(H,27,28)/t25-/m0/s1
LEDER C[C@@](Cc1ccccc1)(NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)O
Molekylvikt (g/mol) 401.45
Formel vikt 401.1627