Läs mer
(Fmoc-Cys-OSu)2, TRC
Produktkod. 30405709
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30405709 500 mg
30405708 50 mg
30404419 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30405709 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF657730500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 112514-60-0
Molekylformel C44 H38 N4 O12 S2
Molekylvikt (g/mol) 878.92
Synonym (3R,8R)-3,8-bis[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-12-(9H-fluoren-9-yl)-10-oxo-11-Oxa-5,6-dithia-2,9-diazadodecanoic acid 9H-fluoren-9-ylmethyl ester, (5R,10R)-5,10-bis[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1-(9H-fluoren-9-yl)-3-oxo-2-Oxa-7,8-dithia-4,11-diazadodecan-12-oic acid 9H-fluoren-9-ylmethyl, [R-(R*,R*)]-5,10-bis[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1-(9H-fluoren-9-yl)-3-oxo-2-Oxa-7,8-dithia-4,11-diazadodecan-12-oic acid 9H-fluoren-9-ylmethyl ester
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) (2R)-3-[[(2R)-3-(2,5-dioxopyrrolidin-1-yl)oxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-oxo-propyl]disulfanyl]-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoate
LEDER O=C(N[C@@H](CSSC[C@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)ON4C(=O)CCC4=O)C(=O)ON5C(=O)CCC5=O)OCC6c7ccccc7c8ccccc68
Specifikationer
Kemiskt namn eller material (Fmoc-Cys-OSu)2
InChI formel InChI=1S/C44H38N4O12S2/c49-37-17-18-38(50)47(37)59-41(53)35(45-43(55)57-21-33-29-13-5-1-9-25(29)26-10-2-6-14-30(26)33)23-61-62-24-36(42(54)60-48-39(51)19-20-40(48)52)46-44(56)58-22-34-31-15-7-3-11-27(31)28-12-4-8-16-32(28)34/h1-16,33-36H,17-24H2,(H,45,55)(H,46,56)/t35-,36-/m0/s1
Kvantitet 500 mg
Formel vikt 878.1928