Läs mer
Fmoc-Dap(Mtt)-OH, TRC
Produktkod. 30179081
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179081 500 mg
30179015 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF628600500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 654670-89-0
Molekylformel C38H34N2O4
Molekylvikt (g/mol) 582.7
Synonym Fmoc-(N-beta-4-methyltrityl)-L-alpha,beta-diaminopropionic Acid, N-α-(9-Fluorenylmethyloxycarbonyl)-N-beta-(p-methyltrityl)-L-2,3-diaminopropionic Acid, (2S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-3-[[(4-methylphenyl)-diphenylmethyl]amino]propanoic Acid
LEDER Cc1ccc(cc1)C(NCC(NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)O)(c5ccccc5)c6ccccc6
Specifikationer
Kemiskt namn eller material Fmoc-Dap(Mtt)-OH
InChI formel InChI=1S/C38H34N2O4/c1-26-20-22-29(23-21-26)38(27-12-4-2-5-13-27,28-14-6-3-7-15-28)39-24-35(36(41)42)40-37(43)44-25-34-32-18-10-8-16-30(32)31-17-9-11-19-33(31)34/h2-23,34-35,39H,24-25H2,1H3,(H,40,43)(H,41,42)/t35-/m0/s1