Läs mer
Fmoc-L-cyclopropylalanine, TRC
Produktkod. 30179076
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179076 1 g
30179134 5 g
30179340 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179076 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF6227801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 214750-76-2
Molekylformel C21 H21 N O4
Molekylvikt (g/mol) 351.4
Synonym Fmoc-β-cyclopropyl-L-Alanine, N-Fmoc-cyclopropyl alanine, N-(9-fluorenylmethoxycarbonyl)-3-cyclopropyl-L-alanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopropylpropanoic Acid
IUPAC-namn (2S)-3-cyclopropyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
LEDER OC(=O)[C@H](CC1CC1)NC(=O)OCC2c3ccccc3c4ccccc24
Specifikationer
Kemiskt namn eller material Fmoc-L-cyclopropylalanine
InChI formel InChI=1S/C21H21NO4/c23-20(24)19(11-13-9-10-13)22-21(25)26-12-18-16-7-3-1-5-14(16)15-6-2-4-8-17(15)18/h1-8,13,18-19H,9-12H2,(H,22,25)(H,23,24)/t19-/m0/s1
Kvantitet 1 g
Formel vikt 351.1471