missing translation for 'onlineSavingsMsg'
Learn More

FMOC-L-Norleucine, 98%, Thermo Scientific Chemicals

Catalog Number p-4323582
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
77284-32-3
C21H23NO4
353.42
MFCD00037537
VCFCFPNRQDANPN-IBGZPJMESA-N
fmoc-nle-oh, fmoc-l-norleucine, n-fmoc-l-norleucine, s-2-fmoc-amino caproic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-norleucine, fmocnleoh, fmoc-nle
7009636
CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
77284-32-3
C21H23NO4
353.42
MFCD00037537
VCFCFPNRQDANPN-IBGZPJMESA-N
fmoc-nle-oh, fmoc-l-norleucine, n-fmoc-l-norleucine, s-2-fmoc-amino caproic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-norleucine, fmocnleoh, fmoc-nle
7009636
CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 138°C to 142°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -19
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
Formula Weight 353.42
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material FMOC-L-Norleucine
Show More Show Less
compliance-icons
Product Identifier
  • FMOC-L-Norleucine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'