Läs mer
Fmoc-L-Tyr(tBu)-OH, TRC
Produktkod. 30179344
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179344 10 g
30179487 5 g
30179055 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179344 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF62286010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71989-38-3
Molekylformel C28H29NO5
Molekylvikt (g/mol) 459.53
Synonym (S)-3-(4-tert-Butoxyphenyl)-2-[[[(9H-fluoren-9-yl)methoxy]carbonyl]amino]propionic Acid, NSC 334301, O-(1,1-Dimethylethyl)-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-tyrosine;
LEDER CC(C)(C)Oc1ccc(C[C@H](NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)O)cc1
Specifikationer
Kemiskt namn eller material Fmoc-L-Tyr(tBu)-OH
InChI formel InChI=1S/C28H29NO5/c1-28(2,3)34-19-14-12-18(13-15-19)16-25(26(30)31)29-27(32)33-17-24-22-10-6-4-8-20(22)21-9-5-7-11-23(21)24/h4-15,24-25H,16-17H2,1-3H3,(H,29,32)(H,30,31)/t25-/m0/s1
Kvantitet 10 g
Formel vikt 459.2046